CH6371378138 Chem Impex International, Inc.

Acetyl-L-threonine methyl ester

  • MDL number MFCD00077085
  • Cas Number : 2458-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Thr-OMe
Purity 95
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/acetyl-l-threonine-methyl-ester/2225?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/acetyl-l-threonine-methyl-ester/2225?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart