CH6371378139 Chem Impex International, Inc.

Na-Acetyl-L-tryptophan 4-nitrophenyl ester

  • MDL number MFCD00037937
  • Cas Number : 14009-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ac-L-Trp-ONp
Purity 98
Storage temp. Store at 0-8
Physical Form Yellowish-greenish powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-acetyl-l-tryptophan-4-nitrophenyl-ester/2226?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-acetyl-l-tryptophan-4-nitrophenyl-ester/2226?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart