CH6371378146 Chem Impex International, Inc.

Ala-Gly-Gly-Gly-OH

  • MDL number MFCD00025559
  • Cas Number : 5123-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at =-4
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/ala-gly-gly-gly-oh/2235?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/ala-gly-gly-gly-oh/2235?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart