CH6371378153 Chem Impex International, Inc.

L-Arginine amide dihydrochloride

  • MDL number MFCD00058286
  • Cas Number : 14975-30-5
  • Molecular Weight : 246.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-arginine-amide-dihydrochloride/2242?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-arginine-amide-dihydrochloride/2242?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart