CH6371378161 Chem Impex International, Inc.

O-tert-Butyl-D-threonine

  • MDL number MFCD00171400
  • Cas Number : 201274-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Thr(tBu)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-d-threonine/2252?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-i-tert-i-butyl-d-threonine/2252?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart