CH6371378171 Chem Impex International, Inc.

L-Glutamic acid gamma-7-amido-4-methylcoumarin

  • MDL number MFCD00038085
  • Cas Number : 72669-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-196 °C (dec.)
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-gamma-7-amido-4-methylcoumarin/2266?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-gamma-7-amido-4-methylcoumarin/2266?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart