CH6371378172 Chem Impex International, Inc.

L-Glutamic acid alpha-4-methoxy-beta-naphthylamide

  • MDL number MFCD00055902
  • Cas Number : 74938-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-186 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form Yellowish powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-4-methoxy-ß-naphthylamide/2267?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-glutamic-acid-a-4-methoxy-ß-naphthylamide/2267?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart