CH6371378178 Chem Impex International, Inc.

Gly-Pro-OH

  • MDL number MFCD00020840
  • Cas Number : 704-15-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Glycyl-L-proline
Purity 99
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/gly-pro-oh/2275?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/gly-pro-oh/2275?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart