CH6371378182 Chem Impex International, Inc.

L-Histidine amide dihydrochloride

  • MDL number MFCD00058559
  • Cas Number : 71666-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-histidine-amide-dihydrochloride/2280?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-histidine-amide-dihydrochloride/2280?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart