CH6371378188 Chem Impex International, Inc.

Leu-Ala-OH

  • MDL number MFCD00002641
  • Cas Number : 7298-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym L-Leucyl-L-alanine
Purity 97
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/leu-ala-oh/2288?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/leu-ala-oh/2288?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart