CH6371378192 Chem Impex International, Inc.

3-(1-Naphthyl)-D-alanine

  • MDL number MFCD00038544
  • Cas Number : 78306-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-3-Ala(1-naphthyl)-OH
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-1-naphthyl-d-alanine/2294?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-1-naphthyl-d-alanine/2294?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart