CH6371378198 Chem Impex International, Inc.

L-Phenylalanine 4-nitroanilide

  • MDL number MFCD00038110
  • Cas Number : 2360-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-155
Synonym L-Phe-pNA
Purity 99
Storage temp. Store at 0-8
Physical Form Light yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-phenylalanine-4-nitroanilide/2302?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-phenylalanine-4-nitroanilide/2302?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart