CH6371378199 Chem Impex International, Inc.

O-Phospho-D-serine

  • MDL number MFCD00069582
  • Cas Number : 73913-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym D-Ser(H2PO3)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-phospho-d-serine/2303?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-phospho-d-serine/2303?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart