CH6371378201 Chem Impex International, Inc.

Pro-Gln-OH

  • MDL number MFCD00037873
  • Cas Number : 18668-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/pro-gln-oh/2305?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/pro-gln-oh/2305?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart