CH6371378206 Chem Impex International, Inc.

L-Tryptophan benzyl ester hydrochloride

  • MDL number MFCD00058122
  • Cas Number : 35858-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216-222� C
Synonym L-Trp-OBzl�
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-tryptophan-benzyl-ester-hydrochloride/2312?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-tryptophan-benzyl-ester-hydrochloride/2312?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart