CH6371378217 Chem Impex International, Inc.

Na,im-Bis-Fmoc-L-histidine

  • MDL number MFCD00070658
  • Cas Number : 98929-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-156
Synonym Fmoc-L-His(Fmoc)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-im-sup-i-i-bis-i-i-fmoc-l-histidine/2373?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-im-sup-i-i-bis-i-i-fmoc-l-histidine/2373?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart