CH6371378234 Chem Impex International, Inc.

5'-O-(4,4'-dimethoxytrityl)-2'-deoxyguanosine

  • MDL number MFCD00063381
  • Cas Number : 81144-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5'-DMT-2'-dG
Purity 98.5
Storage temp. Store at 0-8
Physical Form Off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-2-deoxyguanosine/2397?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/5-i-o-i-4-4-dimethoxytrityl-2-deoxyguanosine/2397?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart