CH6371378241 Chem Impex International, Inc.

Na-Fmoc-Ng-2,4,6-trimethoxybenzyl-L-asparagine

  • MDL number MFCD00153353
  • Cas Number : 120658-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 156-163
Synonym Fmoc-L-Asn(Tmob)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form Off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-2-4-6-trimethoxybenzyl-l-asparagine/2405?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-gamma-sup-2-4-6-trimethoxybenzyl-l-asparagine/2405?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart