CH6371378253 Chem Impex International, Inc.

Boc-O-benzyl-L-threonine N-hydroxysuccinimide ester

  • MDL number MFCD00037905
  • Cas Number : 32886-43-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Thr(Bzl)-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-threonine-i-n-i-hydroxysuccinimide-ester/2438?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-o-i-benzyl-l-threonine-i-n-i-hydroxysuccinimide-ester/2438?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart