CH6371378264 Chem Impex International, Inc.

Boc-beta-(2-thienyl)-D-alanine dicyclohexylammonium salt

  • MDL number MFCD00236874
  • Cas Number : 78452-59-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 227 - 231 °C
Purity 99
Storage temp. Store at 0 - 8 °C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-ß-2-thienyl-d-alanine-dicyclohexylammonium-salt/2449?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-ß-2-thienyl-d-alanine-dicyclohexylammonium-salt/2449?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart