CH6371378273 Chem Impex International, Inc.

Nalpha-Fmoc-Nbeta-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-D-2,3-diaminopropionic acid

  • MDL number MFCD01631975
  • Cas Number : 210830-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-181 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-beta-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-d-2-3-diaminopropionic-acid/2460?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-beta-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-d-2-3-diaminopropionic-acid/2460?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart