CH6371378274 Chem Impex International, Inc.

Fmoc-L-glutamic acid a-9-fluorenylmethyl ester

  • MDL number MFCD00237656
  • Cas Number : 200616-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171-178
Synonym Fmoc-L-Glu-OFm
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-alpha-9-fluorenylmethyl-ester/2461?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-alpha-9-fluorenylmethyl-ester/2461?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart