CH6371378279 Chem Impex International, Inc.

Nalpha-Fmoc-Nim-methyltrityl-L-histidine

  • MDL number MFCD00153358
  • Cas Number : 133367-34-7
  • Molecular Weight : 633.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-145 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystals
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-im-sup-methyltrityl-l-histidine/2466?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-fmoc-i-n-i-sup-im-sup-methyltrityl-l-histidine/2466?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart