CH6371378280 Chem Impex International, Inc.

Na-Fmoc-Nim-Boc-D-histidine cyclohexylammonium salt

  • MDL number MFCD00270617
  • Cas Number : 210755-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-D-His(Boc)-OH CHA
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-im-sup-boc-d-histidine-cyclohexylammonium-salt/2467?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-fmoc-i-n-i-sup-im-sup-boc-d-histidine-cyclohexylammonium-salt/2467?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart