CH6371378282 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-Boc-L-lysine N-hydroxysuccinimide ester

  • MDL number MFCD00153359
  • Cas Number : 132307-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-boc-l-lysine-i-n-i-hydroxysuccinimide-ester/2469?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-e-sup-boc-l-lysine-i-n-i-hydroxysuccinimide-ester/2469?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart