CH6371378287 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-D-lysine

  • MDL number MFCD01862894
  • Cas Number : 333973-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-135 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-d-lysine/2475?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-d-lysine/2475?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart