CH6371378289 Chem Impex International, Inc.

Fmoc-L-methionine sulfone

  • MDL number MFCD00153361
  • Cas Number : 163437-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-171
Synonym Fmoc-L-Met(O2)-OH
Purity 98
Storage temp. Store at RT
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-methionine-sulfone/2477?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-methionine-sulfone/2477?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart