CH6371378293 Chem Impex International, Inc.

Nalpha-Fmoc-Ndelta-methyltrityl-L-Ornithine

  • MDL number MFCD00797872
  • Cas Number : 343770-23-0
  • Molecular Weight : 610.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white crystals or powder
Shipping Temperature RT
Country of Origin US
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-delta-sup-methyltrityl-l-ornithine/2481?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-delta-sup-methyltrityl-l-ornithine/2481?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart