CH6371378294 Chem Impex International, Inc.

Na-Fmoc-Nd-Boc-D-ornithine

  • MDL number MFCD00077065
  • Cas Number : 118476-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133-136
Synonym Fmoc-D-Orn(Boc)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White crystalline powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-d-sup-boc-d-ornithine/2482?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-fmoc-i-n-i-sup-d-sup-boc-d-ornithine/2482?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart