CH6371378300 Chem Impex International, Inc.

Fmoc-O-tert-butyl-L-serine N-hydroxysuccinimide ester

  • MDL number MFCD00153363
  • Cas Number : 158000-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-102 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-i-tert-i-butyl-l-serine-i-n-i-hydroxysuccinimide-ester/2488?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-i-tert-i-butyl-l-serine-i-n-i-hydroxysuccinimide-ester/2488?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart