CH6371378313 Chem Impex International, Inc.

Fmoc-4-(Boc-amino)-L-phenylalanine

  • MDL number MFCD00237007
  • Cas Number : 174132-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Phe(4-NHBoc)-OH
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-4-boc-amino-l-phenylalanine/2505?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-4-boc-amino-l-phenylalanine/2505?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart