CH6371378317 Chem Impex International, Inc.

Fmoc-L-aspartic acid b-9-fluorenylmethyl ester

  • MDL number MFCD01631977
  • Cas Number : 608512-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Asp(OFm)-OH
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-aspartic-acid-b-9-fluorenylmethyl-ester/2509?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-aspartic-acid-b-9-fluorenylmethyl-ester/2509?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart