CH6371378320 Chem Impex International, Inc.

Fmoc-L-glutamic acid g-9-fluorenylmethyl ester

  • MDL number MFCD01631978
  • Cas Number : 608512-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Fmoc-L-Glu(OFm)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-9-fluorenylmethyl-ester/2512?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-glutamic-acid-9-fluorenylmethyl-ester/2512?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart