CH6371378325 Chem Impex International, Inc.

Fmoc-O-acetyl-D-serine

  • MDL number MFCD00077069
  • Cas Number : 608512-87-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-165� C
Synonym Fmoc-D-Ser(Ac)-OH
Purity 97
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-acetyl-d-serine/2517?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-o-i-acetyl-d-serine/2517?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart