CH6371378336 Chem Impex International, Inc.

Boc-L-glutamic acid a-amide

  • MDL number MFCD00070081
  • Cas Number : 18800-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Glu-NH2
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-a-amide/2529?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-a-amide/2529?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart