CH6371378339 Chem Impex International, Inc.

Boc-L-glutamic acid g-tert-butyl ester a-N-hydroxysuccinimide ester

  • MDL number MFCD00153303
  • Cas Number : 32886-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-145
Synonym Boc-L-Glu(OtBu)-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-i-tert-i-butyl-ester-a-i-n-i-hydroxysuccinimide-ester/2532?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamic-acid-i-tert-i-butyl-ester-a-i-n-i-hydroxysuccinimide-ester/2532?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart