CH6371378340 Chem Impex International, Inc.

Boc-L-glutamine N-hydrosuccinimide ester

  • MDL number MFCD00055962
  • Cas Number : 18800-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Gln-OSu
Purity 98
Storage temp. Store at = -15
Physical Form White to off-white powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-glutamine-i-n-i-hydrosuccinimide-ester/2533?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-glutamine-i-n-i-hydrosuccinimide-ester/2533?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart