CH6371378342 Chem Impex International, Inc.

Boc-glycine 4-nitrophenyl ester

  • MDL number MFCD00038125
  • Cas Number : 3655-05-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-66
Synonym Boc-Gly-ONp
Purity 98
Storage temp. Store at 0-8
Physical Form Brown powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-glycine-4-nitrophenyl-ester/2536?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-glycine-4-nitrophenyl-ester/2536?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart