CH6371378345 Chem Impex International, Inc.

Boc-L-homocitruline

  • MDL number MFCD00151903
  • Cas Number : 201418-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-HomoCit-OH
Purity 98
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-homocitruline/2539?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-homocitruline/2539?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart