CH6371378352 Chem Impex International, Inc.

Boc-L-phenylalanine N-methoxy-N-methyl amide

  • MDL number MFCD00151891
  • Cas Number : 87694-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Phe-N(OCH3)CH3
Purity 98
Storage temp. Store at 0-8
Physical Form White to off-white solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-phenylalanine-i-n-i-methoxy-i-n-i-methyl-amide/2547?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-phenylalanine-i-n-i-methoxy-i-n-i-methyl-amide/2547?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart