CH6371378355 Chem Impex International, Inc.

Boc-N-methyl-D-phenylalanine dicyclohexylammonium salt

  • MDL number MFCD00058079
  • Cas Number : 102185-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174-178� C
Synonym Boc-N-Me-D-Phe-OH�A
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-d-phenylalanine-dicyclohexylammonium-salt/2550?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methyl-d-phenylalanine-dicyclohexylammonium-salt/2550?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart