CH6371378356 Chem Impex International, Inc.

Boc-L-proline 4-nitrophenyl ester

  • MDL number MFCD00037323
  • Cas Number : 28310-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-66 �C
Synonym Boc-L-Pro-ONp
Purity 99
Storage temp. Store at 0-8
Physical Form Pale beige powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-proline-4-nitrophenyl-ester/2551?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-proline-4-nitrophenyl-ester/2551?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart