CH6371378357 Chem Impex International, Inc.

Boc-D-proline N-hydroxysuccinimide ester

  • MDL number MFCD00069687
  • Cas Number : 102185-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-137
Synonym Boc-D-Pro-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-proline-i-n-i-hydroxysuccinimide-ester/2552?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-proline-i-n-i-hydroxysuccinimide-ester/2552?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart