CH6371378358 Chem Impex International, Inc.

Boc-N-methylglycine N-hydroxysuccinimide ester

  • MDL number MFCD00153312
  • Cas Number : 80621-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-131 °C
Purity 98
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-n-i-methylglycine-i-n-i-hydroxysuccinimide-ester/2553?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-n-i-methylglycine-i-n-i-hydroxysuccinimide-ester/2553?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart