CH6371378361 Chem Impex International, Inc.

Na-Boc-L-tryptophan 4-nitrophenyl ester

  • MDL number MFCD00022757
  • Cas Number : 15160-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Trp-ONp
Purity 98
Storage temp. Store at 0-8
Physical Form Yellow powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-tryptophan-4-nitrophenyl-ester/2556?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-l-tryptophan-4-nitrophenyl-ester/2556?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart