CH6371378362 Chem Impex International, Inc.

Na-Boc-L-tryptophan N-methoxy-N-methyl amide

  • MDL number MFCD00153482
  • Cas Number : 97530-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Trp-N(OCH3)CH3
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-l-tryptophan-i-n-i-methoxy-i-n-i-methyl-amide/2557?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-sup-i-i-boc-l-tryptophan-i-n-i-methoxy-i-n-i-methyl-amide/2557?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart