CH6371378364 Chem Impex International, Inc.

Boc-L-tyrosine N-hydroxysuccinimide ester

  • MDL number MFCD00037910
  • Cas Number : 20866-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Tyr-OSu
Purity 98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-l-tyrosine-i-n-i-hydroxysuccinimide-ester/2559?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-l-tyrosine-i-n-i-hydroxysuccinimide-ester/2559?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart