CH6371378368 Chem Impex International, Inc.

Boc-S-3-nitro-2-pyridinesulfenyl-L-cysteine

  • MDL number MFCD00153301
  • Cas Number : 76880-29-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Boc-L-Cys(Npys)-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-nitro-2-pyridinesulfenyl-l-cysteine/2563?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-nitro-2-pyridinesulfenyl-l-cysteine/2563?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart