CH6371378373 Chem Impex International, Inc.

Z-g-aminobutyric acid

  • MDL number MFCD00055835
  • Cas Number : 5105-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Z-gamma-Abu-OH
Purity 99
Storage temp. Store at 0-8
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-aminobutyric-acid/2569?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-aminobutyric-acid/2569?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart