CH6371378375 Chem Impex International, Inc.

Na-Z-Nw-(2,2,5,7,8-pentamethylchroman-6-sulfonyl)-L-arginine cyclohexylammonium salt

  • MDL number MFCD00153323
  • Cas Number : 112160-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158-170
Synonym Z-L-Arg(Pmc)-OH CHA
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-omega-sup-2-2-5-7-8-pentamethylchroman-6-sulfonyl-l-arginine-cyclohexylammonium-salt/2571?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-z-i-n-i-sup-omega-sup-2-2-5-7-8-pentamethylchroman-6-sulfonyl-l-arginine-cyclohexylammonium-salt/2571?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart